Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium 1-butanesulfonate, For ion pair chromatography, 99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013647 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3647 50 g
11-101-3646 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3647 Supplier MilliporeSigma Supelco Supplier No. 1902250GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2386-54-1 | Formula: C4H9NaO3S | Weight: 160.16 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym 1-Butanesulfonic acid sodium salt
IUPAC Name sodium butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Physical Form Solid
Quantity 50 g
Product Line LiChropur
Linear Formula CH3(CH2)3 SO3Na
Formula Weight 160.17
Percent Purity ≥99.0% (T)
Grade Ion Pair Chromatography
Chemical Name or Material Sodium 1-butanesulfonate

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.