Learn More
LiChropur™ Sodium 1-decanesulfonate ,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Melting Point | >300°C |
| Physical Form | Solid |
| Quantity | 50 g |
| Linear Formula | CH3(CH2)8 CH2 SO3 Na |
| Formula Weight | 244.33 |
| Percent Purity | ≥99.0% (T); ≥99.0% |
| Grade | Ion Pair Chromatography |
| Product Line | LiChropur |
| Chemical Name or Material | Sodium 1-decanesulfonate |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.