Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium 1-heptanesulfonate monohydrate, 99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013497 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3497 50 g
11-101-3495 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3497 Supplier MilliporeSigma Supelco Supplier No. 5183250GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Description Anionic
CAS 207300-90-1
Product Line LiChropur
Linear Formula CH3(CH2)5 CH2 SO3 Na · H2O
MDL Number MFCD00149550
Synonym 1-Heptanesulfonic acid sodium salt monohydrate
Type Organic Standard
Quantity 50 g
Formula Weight 220.26
Percent Purity ≥99% (T)
Grade Ion Pair Chromatography
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.