Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium 1-Hexanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013499 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3499 10 g
11-101-3500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3499 Supplier MilliporeSigma Supelco Supplier No. 5286210GF
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00149549
Synonym 1-Hexanesulfonic acid sodium salt monohydrate

Specifications

CAS 207300-91-2
Linear Formula CH3(CH2)4 CH2 SO3 Na · H2O
Quantity 10 g
Formula Weight 206.24
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.