Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium 1-propanesulfonate monohydrate , 98.0-102.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013868 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3868 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3868 Supplier MilliporeSigma Supelco Supplier No. 8180610GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00167033
Synonym 1-Propanesulfonic acid sodium salt monohydrate

Specifications

Melting Point 250°C
CAS 304672-01-3
Linear Formula CH3 CH2 CH2 SO3 Na · H2O
Quantity 10 g
Formula Weight 164.16
Percent Purity 98.0 to 102.0% (T)
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.