Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium dodecyl sulfate,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111019029 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9029 10 g
11-101-8112 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9029 Supplier MilliporeSigma Supelco Supplier No. 7172610GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium dodecyl sulfate (SDS) is a detergent suitable to denature proteins. It is a long-chain aliphatic group which has negatively charged sulfate, hence making it amphipathic detergent in nature.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00036175
Synonym Dodecyl sodium sulfate; Dodecyl sulfate sodium salt; Lauryl sulfate sodium salt; SDS; Sodium lauryl sulfate

Specifications

Melting Point 204°C to 207°C
CAS 151-21-3
Physical Form Rod
Quantity 10 g
Linear Formula CH3(CH2)11 OSO3 Na
Flash Point 170°C
Solubility Information Water: soluble
Molecular Weight (g/mol) 288.38
Formula Weight 288.38
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Ion Pair Chromatography
Product Line LiChropur
Chemical Name or Material Sodium Dodecyl Sulfate
Show More Show Less
RTECSNumber WT1050000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.