Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Tetrabutylammonium bisulfate,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013925 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
10 x 1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3925 10 x 1 g
11-101-3926 10 g
11-101-3927 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-3925 Supplier MilliporeSigma Supelco Supplier No. 8685310X1GF
Add to Cart
Add to Cart

Tetrabutyammonium bisulfate (TBAHS) is an ion-pairing reagent. It acts as a catalyst in hydrolysis of aziridines and epoxides and the catalyst can be recycled.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011637
Synonym Tetrabutylammonium hydrogen sulfate

Specifications

Melting Point 171°C to 174°C
CAS 32503-27-8
Flash Point 175.6°C - Closed Cup
Linear Formula (CH3 CH2 CH2 CH2)4 N(HSO4)
Quantity 10 x 1 g
Formula Weight 339.53
Percent Purity ≥99.0% (T); ≥99.0%
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.