Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Tetrahexylammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111014198 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4198 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4198 Supplier MilliporeSigma Supelco Supplier No. 8729710GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity ion-pairing reagent designed for optimized reversed-phase HPLC and LC/MS applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 434.58
MDL Number MFCD00011858

Specifications

Melting Point 99°C to 100°C
CAS 4328-13-6
Linear Formula [CH3(CH2)5]4 N(Br)
Quantity 10 g
Formula Weight 434.58
Percent Purity ≥99.0% (AT)
Grade For ion pair chromatography
Physical Form Crystals
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.