Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Tetrapropylammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111013431 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3431 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3431 Supplier MilliporeSigma Supelco Supplier No. 8810310G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mobile phase additive for ion-pair chromatography and HPLC analysis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 266.26
MDL Number MFCD00011840

Specifications

CAS 1941-30-6
Linear Formula (CH3 CH2 CH2)4 N(Br)
Quantity 10 g
Formula Weight 266.26
Percent Purity ≥99.0% (AT)
Grade For ion pair chromatography
Physical Form Crystals
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.