Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Triethylamine Hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111022700 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2700 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2700 Supplier MilliporeSigma Supelco Supplier No. 6396150G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-68-7
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00012500
InChI Key ILWRPSCZWQJDMK-UHFFFAOYSA-N
IUPAC Name hydrogen triethylamine chloride
SMILES [H+].[Cl-].CCN(CC)CC

Specifications

Melting Point 256°C to 259°C; 261°C (decomposition) (literature)
Linear Formula (C2H5)3 N · HCl
Quantity 50 g
Formula Weight 137.65
Percent Purity 99% to 101% (AT)
Grade For HPLC
Physical Form Solid
Hazard Category H314
RTECSNumber YE2111500

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.