Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Triethylamine, MilliporeSigma™ Supelco™

Catalog No. 111022786 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2786 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2786 Supplier MilliporeSigma Supelco Supplier No. 8110110X2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 101.19

Specifications

Melting Point -115°C (literature)
pH 12.7 (15°C, 100 g/L)
Quantity 10 x 2 mL
Formula Weight 101.19
Percent Purity ≥99.5% (GC)
Grade HPLC
Physical Form Liquid

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.