Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Triethylammonium bicarbonate buffer, 1 M, MilliporeSigma™ Supelco™

Catalog No. 111013260 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3260 100 mL
11-101-3261 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3260 Supplier MilliporeSigma Supelco Supplier No. 18597100ML
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00466938
Synonym Triethylammonium hydrogen carbonate buffer

Specifications

CAS 15715-58-9
Density 1.015-1.024
pH 8.4 to 8.6 (20°C in neat)
Quantity 100 mL
Grade For HPLC
Concentration or Composition (by Analyte or Components) 1 M
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.