Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Trifluoroacetic Anhydride, For GC derivatization, 99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018735 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8735 10 x 1 mL
11-101-9765 10 mL
11-101-8363 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8735 Supplier MilliporeSigma Supelco Supplier No. 9171910X1ML
Add to Cart
Add to Cart

Reagent for cabethoxy, butyl ester, n-methyl, methyl ester, di(trifluoroacetyl), hexafluoroisopropyl ester, acetyl or trifluoroacetyl phenylthihydantoins, trifluoroacetyl (2-hydroxy-2-methyl)ethyl, n-trifluoroacetyl, trifluoroacetyl (TFA), trifluoroacetamide, 4-trifluoromethyl-2, 6-diamino-1,3,5-triazineacetyl and trifluoroacetyl vanillin.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym TFAA
IUPAC Name 2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F

Specifications

Melting Point -65°C
Boiling Point 39.5°C to 40°C
Linear Formula (CF3CO)2 O
Formula Weight 210.03
Percent Purity ≥99.0% (GC)
Grade GC/MS (SIM)
Density 1.511 g/mL (at 20°C)
Quantity 10 x 1 mL
Refractive Index n20/D 1.3
Vapor Pressure 6.28 psi (20°C)
RTECSNumber AJ9800000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.