Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Trimethylphenylammonium hydroxide Solution, ∼0.5M (CH3)3N(OH)C6H5 in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111019302 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9302 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9302 Supplier MilliporeSigma Supelco Supplier No. 7926610X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trimethylphenylammonium hydroxide (TMAH) is a methylating reagent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00041899
Synonym Phenyltrimethylammonium hydroxide; TMAH

Specifications

CAS 1899-02-1
Linear Formula (CH3)3 N(OH)C6H5
Quantity 10 x 1 mL
Formula Weight 153.22
Grade For GC derivatization
Concentration or Composition (by Analyte or Components) 0.5 M (CH3)3 N(OH)C6H5 in. methanol
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.