Learn More
Sigma Aldrich Fine Chemicals Biosciences LiChrospherR RP 18 HPLC Co
Catalog No. 502909046
Encompass
LiChrospher(R) RP-18 HPLC column is an incredibly reliable and highly versatile, traditionally manufactured spherical silica-based support, designed for reversed-phase chromatography of acidic, neutral and weakly basic compounds - substances frequently found in all analytical fields. The optimal selectivity and performance of the column ensure that these parameters remain constant from batch to batch and from year to year.
Catalog No. 50-290-9046 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 50139U
LiChrospher(R) RP-18 HPLC column is an incredibly reliable and highly versatile, traditionally manufactured spherical silica-based support, designed for reversed-phase chromatography of acidic, neutral and weakly basic compounds - substances frequently found in all analytical fields. The optimal selectivity and performance of the column ensure that these parameters remain constant from batch to batch and from year to year.