Learn More
Sigma Aldrich Fine Chemicals Biosciences LiChrospherR RP Select B H
Catalog No. 502908504
Encompass
LiChrospher(R) RP-select B HPLC column is a versatile reversed-phase sorbent optimized for HPLC separations of basic compounds. Based on spherical silica particles, the sorbent prevents secondary interactions with basic substances, and ensures that they are eluted as highly symmetrical peaks. It is also suitable for the determination of neutral and acidic substances.
Catalog No. 50-290-8504 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 50146U
LiChrospher(R) RP-select B HPLC column is a versatile reversed-phase sorbent optimized for HPLC separations of basic compounds. Based on spherical silica particles, the sorbent prevents secondary interactions with basic substances, and ensures that they are eluted as highly symmetrical peaks. It is also suitable for the determination of neutral and acidic substances.