Learn More
Apexbio Technology LLC LiCl(Synonyms: Lithium chloride, Lithium(I) chloride, Lithium monochloride, Lithium muriate, LiCl salt), 10mM (in 1mL H2O), CAS: 7447-41-8.
SDP
Catalog No. 5000061006
Encompass_Preferred
Lithium chloride (LiCl CAS 7447-41-8) is an ionic compound widely utilized as a selective precipitant for RNA isolation in molecular biology research Due to its physicochemical properties LiCl preferentially precipitates RNA while generally not interacting appreciably with DNA proteins or polysaccharides Researchers commonly employ LiCl precipitation to purify intact translationally active RNA from complex samples including those containing translation inhibitors or synthesized cDNA Additionally LiCl-mediated RNA precipitation following guanidine thiocyanate homogenization enables rapid scalable purification of large mRNA species up to 10 kb benefiting workflows by avoiding ultracentrifugation steps
Catalog No. 50-000-61006 Supplier Apexbio Technology LLC Supplier No. B60835.1
Lithium chloride (LiCl CAS 7447-41-8) is an ionic compound widely utilized as a selective precipitant for RNA isolation in molecular biology research Due to its physicochemical properties LiCl preferentially precipitates RNA while generally not interacting appreciably with DNA proteins or polysaccharides Researchers commonly employ LiCl precipitation to purify intact translationally active RNA from complex samples including those containing translation inhibitors or synthesized cDNA Additionally LiCl-mediated RNA precipitation following guanidine thiocyanate homogenization enables rapid scalable purification of large mRNA species up to 10 kb benefiting workflows by avoiding ultracentrifugation steps