Learn More
Cayman Chemical Licochalcone A, Purity: ≥95%, CAS: 58749-22-7, 50 mg
SDP
Catalog No. 502953134
Encompass_Preferred
A natural chalcone that has anti-inflammatory actions, supporting its use as a skin lightening agent, particularly for rosacea and atopic dermatitis; induces apoptosis and suppresses migration and invasion of cancer cells in vitro; has antibacterial and anti-parasitic actions
Catalog No. 50-295-3134 Supplier Cayman Chemical Supplier No. 1185350 MG
A natural chalcone that has anti-inflammatory actions, supporting its use as a skin lightening agent, particularly for rosacea and atopic dermatitis; induces apoptosis and suppresses migration and invasion of cancer cells in vitro; has antibacterial and anti-parasitic actions