Learn More
TARGETMOL CHEMICALS INC Licochalcone A 20MG
SDP
Catalog No. 502876455
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. Licochalcone A is a flavonoid isolated from the famous Chinese medicinal herb Glycyrrhiza uralensis Fisch with obvious anti-cancer effects. Purity 98.96%
Catalog No. 50-287-6455 Supplier TARGETMOL CHEMICALS INC Supplier No. T360020MG
Also available in 1 mL and bulk. Please contact Fisher for quotes. Licochalcone A is a flavonoid isolated from the famous Chinese medicinal herb Glycyrrhiza uralensis Fisch with obvious anti-cancer effects. Purity 98.96%