Learn More
Selleck Chemical LLC Licochalcone A
SDP
Supplier:  Selleck Chemical LLC S782825MG
Encompass_Preferred
Licochalcone A is an estrogenic flavanoid extracted from licorice root showing antimalarial anticancer antibacterial and antiviral activities Phase 3
Catalog No. 50-000-00712