Learn More
Apexbio Technology LLC Licochalcone A 58749-22-7 20mg
SDP
Catalog No. 5000094446
Encompass_Preferred
Licochalcone A (CAS 58749-22-7) is a flavonoid compound isolated from the root of Glycyrrhiza species (licorice) It modulates multiple biological pathways including the inhibition of inflammatory mediators such as NF- B and the suppression of oxidative stress by regulating antioxidant enzymes Licochalcone A has demonstrated antiproliferative effects in various cell models impacting cell cycle progression and apoptosis Due to these properties it is widely utilized in studies related to inflammation cancer and oxidative stress-associated diseases The compound should be stored in a cool dry and sealed environment to maintain stability
Catalog No. 50-000-94446 Supplier Apexbio Technology LLC Supplier No. N241720
Licochalcone A (CAS 58749-22-7) is a flavonoid compound isolated from the root of Glycyrrhiza species (licorice) It modulates multiple biological pathways including the inhibition of inflammatory mediators such as NF- B and the suppression of oxidative stress by regulating antioxidant enzymes Licochalcone A has demonstrated antiproliferative effects in various cell models impacting cell cycle progression and apoptosis Due to these properties it is widely utilized in studies related to inflammation cancer and oxidative stress-associated diseases The compound should be stored in a cool dry and sealed environment to maintain stability