Learn More
Medchemexpress LLC Licochalcone C | 144506-14-9 | 99.6% | 338.40 g/mol | C21H22O4 | 10 MG
SDP
Catalog No. 5000295489
Encompass_Preferred
Licochalcone C is a chalcone-derived small molecule provided for research use. It is reported to inhibit α-glucosidase with sub-100 nM potency and to inhibit protein tyrosine phosphatase 1B at micromolar concentrations. Supplied as a high-purity solid suitable for biochemical and enzymatic assays.
  • High purity (99.6%).
  • Molecular formula C21H22O4; molecular weight 338.40 g/mol.
  • Available in small-mass research quantities suitable for assay development.
  • Demonstrated α-glucosidase inhibitory activity (IC50 <100 nM).
  • For research use only; not for human or therapeutic use.
Catalog No. 50-002-95489 Supplier Medchemexpress LLC Supplier No. HYN037410MG
Licochalcone C is a chalcone-derived small molecule provided for research use. It is reported to inhibit α-glucosidase with sub-100 nM potency and to inhibit protein tyrosine phosphatase 1B at micromolar concentrations. Supplied as a high-purity solid suitable for biochemical and enzymatic assays.
  • High purity (99.6%).
  • Molecular formula C21H22O4; molecular weight 338.40 g/mol.
  • Available in small-mass research quantities suitable for assay development.
  • Demonstrated α-glucosidase inhibitory activity (IC50 <100 nM).
  • For research use only; not for human or therapeutic use.