Learn More
Medchemexpress LLC HY-N0374 5mg Medchemexpress, Licochalcone C CAS:144506-14-9 Purity:>98%
SDP
Catalog No. 501970262
Encompass_Preferred
Medchemexpress, HY-N0374 5mg Licochalcone C CAS:144506-14-9 Licochalcone C could inhibit α-glucosidase, with IC50s of <100 nM and 92.43 μM for α-glucosidase and protein tyrosine phosphatase 1B (PTP1B), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0262 Supplier Medchemexpress LLC Supplier No. HYN03745MG
Medchemexpress, HY-N0374 5mg Licochalcone C CAS:144506-14-9 Licochalcone C could inhibit α-glucosidase, with IC50s of <100 nM and 92.43 μM for α-glucosidase and protein tyrosine phosphatase 1B (PTP1B), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.