Learn More
Medchemexpress LLC Licochalcone D | 144506-15-0 | MFCD32197344 | 1 ML
SDP
Catalog No. 5000420804
Encompass_Preferred
Licochalcone D is a flavonoid compound primarily found in the root of *Glycyrrhiza uralensis*. It acts as a potent and orally active inhibitor of NF-κB p65 and exhibits antioxidant, anti-inflammatory, and anti-cancer properties.
  • Derived from the root of Glycyrrhiza uralensis.
  • Potent and orally active inhibitor of NF-κB p65.
  • Possesses antioxidant, anti-inflammatory, and anti-cancer characteristics.
  • Purity: 99.80%.
Catalog No. 50-004-20804 Supplier Medchemexpress LLC Supplier No. HYN418710MM/1ML
Licochalcone D is a flavonoid compound primarily found in the root of *Glycyrrhiza uralensis*. It acts as a potent and orally active inhibitor of NF-κB p65 and exhibits antioxidant, anti-inflammatory, and anti-cancer properties.
  • Derived from the root of Glycyrrhiza uralensis.
  • Potent and orally active inhibitor of NF-κB p65.
  • Possesses antioxidant, anti-inflammatory, and anti-cancer characteristics.
  • Purity: 99.80%.