Learn More
Selleck Chemical LLC Licochalcone D
SDP
Catalog No. 5000002424
Encompass_Preferred
Licochalcone D (Lico D LCD LD) a flavonoid isolated from a Chinese medicinal plant Glycyrrhiza inflata has antioxidant anti-inflammatory and anti-cancer properties Licochalcone D inhibit phosphorylation of NF- B p65 in LPS signaling pathway Licochalcone D inhibits JAK2 EGFR and Met (c-Met) activities and induces ROS-dependent apoptosis Licochalcone D also induces caspases activation and poly (ADP-ribose) polymerase (PARP) cleavage
Catalog No. 50-000-02424 Supplier Selleck Chemical LLC Supplier No. S68991MG
Licochalcone D (Lico D LCD LD) a flavonoid isolated from a Chinese medicinal plant Glycyrrhiza inflata has antioxidant anti-inflammatory and anti-cancer properties Licochalcone D inhibit phosphorylation of NF- B p65 in LPS signaling pathway Licochalcone D inhibits JAK2 EGFR and Met (c-Met) activities and induces ROS-dependent apoptosis Licochalcone D also induces caspases activation and poly (ADP-ribose) polymerase (PARP) cleavage