Learn More
TARGETMOL CHEMICALS INC LICOCHALCONE D 5MG
SDP
Catalog No. 502579873
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Licochalcone D may be a potential drug for human melanoma treatment by inhibiting proliferation inducing apoptosis via the mitochondrial pathway and blocking cell migration and invasion. Licochalcone D has cardioprotective potential against myocardial ischemia/reperfusion injury in langendorff-perfused rat hearts. Licochalcone D has anti-inflammatory activity it shows suppression ability of nitric oxide (NO) production. Licochalcone D has anti-allergic activity it suppresses degranulation by decreasing the intracellular Ca2+ level and tyrosine phosphorylation of ERK in RBL-2H3 cells. purity: 99%
Catalog No. 50-257-9873 Supplier TARGETMOL CHEMICALS INC Supplier No. T45185MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Licochalcone D may be a potential drug for human melanoma treatment by inhibiting proliferation inducing apoptosis via the mitochondrial pathway and blocking cell migration and invasion. Licochalcone D has cardioprotective potential against myocardial ischemia/reperfusion injury in langendorff-perfused rat hearts. Licochalcone D has anti-inflammatory activity it shows suppression ability of nitric oxide (NO) production. Licochalcone D has anti-allergic activity it suppresses degranulation by decreasing the intracellular Ca2+ level and tyrosine phosphorylation of ERK in RBL-2H3 cells. purity: 99%