Learn More
Medchemexpress LLC HY-N4187 5mg Medchemexpress, Licochalcone D CAS:144506-15-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41875MG
Encompass_Preferred
Medchemexpress, HY-N4187 5mg Licochalcone D CAS:144506-15-0 Licochalcone D, a flavonoid compound mainly existing in the root of Glycyrrhiza inflate, is a potent inhibitor of NF-kappaB (NF-κB) p65. Licochalcone D possesses antioxidant, anti-inflammatory, anti-cancer properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0434