Learn More
Medchemexpress LLC HY-N4182 5mg Medchemexpress, Licochalcone E CAS:864232-34-8 Purity:>98%
SDP
Catalog No. 501970432
Encompass_Preferred
Medchemexpress, HY-N4182 5mg Licochalcone E CAS:864232-34-8 Licochalcone E, a flavonoid compound isolated from Glycyrrhiza inflate, inhibits NF-κB and AP-1 transcriptional activity through the inhibition of AKT and MAPK activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0432 Supplier Medchemexpress LLC Supplier No. HYN41825MG
Medchemexpress, HY-N4182 5mg Licochalcone E CAS:864232-34-8 Licochalcone E, a flavonoid compound isolated from Glycyrrhiza inflate, inhibits NF-κB and AP-1 transcriptional activity through the inhibition of AKT and MAPK activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.