Learn More
Medchemexpress LLC HY-B1452 5mg , Licofelone CAS:156897-06-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB14525MG
Encompass_Preferred
HY-B1452 5mg Licofelone CAS: 156897-06-2 Licofelone (ML-3000) is a dual COX/5-lipoxygenase (5-LOX) inhibitor (IC50=0.21/0.18 μM, respectively) for the treatment of osteoarthritis. Licofelone exerts anti-inflammatory and anti-proliferative effects. Licofelone induces apoptosis, and decreases the production of proinflammatory leukotrienes and prostaglandins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1535