Learn More
Medchemexpress LLC Licoisoflavone A | 66056-19-7 | 1 ML
SDP
Catalog No. 5000420027
Encompass_Preferred
Licoisoflavone A is an orally active isoflavone that inhibits proliferation, induces apoptosis, and causes G1/S phase arrest in colorectal cancer (CRC) cells by inhibiting the CDK2-Cyclin E1 axis. It demonstrates a dose-dependent inhibitory effect on SARS-CoV-2 infection and significant anti-tumor efficacy in mice, making it useful for research into colorectal cancer and SARS-CoV-2.
  • Orally active isoflavone
  • Inhibits proliferation, induces apoptosis, and causes G1/S phase arrest in colorectal cancer cells
  • Inhibits the CDK2-Cyclin E1 axis
  • Inhibits lipid peroxidation (IC50 of 7.2 μM)
  • Shows dose-dependent inhibition of SARS-CoV-2 infection
  • Exhibits significant anti-tumor efficacy in mice
Catalog No. 50-004-20027 Supplier Medchemexpress LLC Supplier No. HYN338910MM/1ML
Licoisoflavone A is an orally active isoflavone that inhibits proliferation, induces apoptosis, and causes G1/S phase arrest in colorectal cancer (CRC) cells by inhibiting the CDK2-Cyclin E1 axis. It demonstrates a dose-dependent inhibitory effect on SARS-CoV-2 infection and significant anti-tumor efficacy in mice, making it useful for research into colorectal cancer and SARS-CoV-2.
  • Orally active isoflavone
  • Inhibits proliferation, induces apoptosis, and causes G1/S phase arrest in colorectal cancer cells
  • Inhibits the CDK2-Cyclin E1 axis
  • Inhibits lipid peroxidation (IC50 of 7.2 μM)
  • Shows dose-dependent inhibition of SARS-CoV-2 infection
  • Exhibits significant anti-tumor efficacy in mice