Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Licorice-saponin H2 | 118441-85-3 | 10 MG
SDP

Catalog No. 5000419196
Encompass_Preferred

Licorice-saponin H2 is a triterpenoid oligosaccharide glycoside found in the Glycyrrhiza genus. It accumulates in the roots of licorice plants under salt stress conditions, suggesting a role in plant stress resistance.

  • Purity of 99.72%
  • Molecular weight of 822.93
  • Molecular formula C42H62O16
  • CAS number 118441-85-3
  • White to off-white solid appearance
  • Sourced from Glycyrrhiza uralensis Fisch.
  • Soluble up to 100 mg/mL in DMSO

Catalog No. 50-004-19196 Supplier Medchemexpress LLC Supplier No. HYN691110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Licorice-saponin H2 is a triterpenoid oligosaccharide glycoside found in the Glycyrrhiza genus. It accumulates in the roots of licorice plants under salt stress conditions, suggesting a role in plant stress resistance.

  • Purity of 99.72%
  • Molecular weight of 822.93
  • Molecular formula C42H62O16
  • CAS number 118441-85-3
  • White to off-white solid appearance
  • Sourced from Glycyrrhiza uralensis Fisch.
  • Soluble up to 100 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.