Learn More
Medchemexpress LLC Licoricone | 51847-92-8 | 1 MG
SDP
Catalog No. 5000417764
Encompass_Preferred
Licoricone is a flavonoid extracted from licorice. It demonstrates anti-helicobacter pylori activity against CLAR and AMOX-resistant strains, as well as four CLAR (AMOX)-sensitive strains.
  • Flavonoid extracted from licorice
  • Demonstrates anti-Helicobacter pylori activity
  • Effective against CLAR and AMOX-resistant strains
  • Effective against CLAR (AMOX)-sensitive strains
Catalog No. 50-004-17764 Supplier Medchemexpress LLC Supplier No. HYN33861MG
Licoricone is a flavonoid extracted from licorice. It demonstrates anti-helicobacter pylori activity against CLAR and AMOX-resistant strains, as well as four CLAR (AMOX)-sensitive strains.
  • Flavonoid extracted from licorice
  • Demonstrates anti-Helicobacter pylori activity
  • Effective against CLAR and AMOX-resistant strains
  • Effective against CLAR (AMOX)-sensitive strains