Learn More
Greiner Bio-One Lid for 24-well Microplates
GREENER_CHOICE
Catalog No. 07000671 Shop All Greiner Bio-One Products
Change view
Click to view available options
Plate Compatibility:
24-well Microplates
Catalog No. Plate Compatibility
07-000-671 24-well Microplates
1 options
Catalog No. 07-000-671 Supplier Greiner Bio-One Supplier No. 662124
Add to Cart
Add to Cart
Lid for 24-Well Microplates.
  • Made of polystyrene
  • Features condensation rings for tight seal
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Sterility Non-sterile
No. of Wells 24
Condensation Rings Yes
Plate Compatibility 24-well Microplates