Learn More
Akro Mils Inc Clear snap-on lids for Akro-Mils Akro-Grid Dividable Containers
Catalog No. NC2118717
Encompass_Preferred
This item must be ordered in increments of 3 per supplier requirements
Clear snap-on lids for Akro-Mils Akro-Grid Dividable Containers. Lids snap securely to the rim of grid boxes to keep contents clean yet visible. Containers stack with lids in place.AKRO MILS INC, mfg# 33023
This item is not returnable. View return policy
Catalog No. NC2118717 Supplier Akro Mils Inc Supplier No. 33023
This item must be ordered in increments of 3 per supplier requirements
Clear snap-on lids for Akro-Mils Akro-Grid Dividable Containers. Lids snap securely to the rim of grid boxes to keep contents clean yet visible. Containers stack with lids in place.AKRO MILS INC, mfg# 33023