Learn More
Electron Microscopy Sciences StainTray™ Black Lid Only for 10 Slide Stain Tray
SDP
Catalog No. 501899565
Encompass_Preferred
A black cover for fluorescent work, made from ABS with a temperature range of -80ºC to 80ºC.
Capacity: 10 slides
Catalog No. 50-189-9565 Supplier Electron Microscopy Sciences Supplier No. 71398B
A black cover for fluorescent work, made from ABS with a temperature range of -80ºC to 80ºC.
Capacity: 10 slides