Learn More
Electron Microscopy Sciences MicroWell Lid for 96-Well Plate
SDP
Catalog No. 50930291
Encompass
Vented lids for MicroWell plates allow for gas exchange and have condensation rings to control evaporation and retain condensate.
Catalog No. 50-930-291 Supplier Electron Microscopy Sciences Supplier No. 70436L5
Vented lids for MicroWell plates allow for gas exchange and have condensation rings to control evaporation and retain condensate.