Learn More
Electron Microscopy Sciences MicroWell Lid for 96-Well Plate
SDP
Catalog No. 50930290
Encompass
Vented lids for MicroWell plates allow for gas exchange and have condensation rings to control evaporation and retain condensate.
Catalog No. 50-930-290 Supplier Electron Microscopy Sciences Supplier No. 70436L1
Vented lids for MicroWell plates allow for gas exchange and have condensation rings to control evaporation and retain condensate.