Learn More
Research Products International Corp Plastic Lid for Glass Staining Dish, Large, 38.6 x 26.5 x 4.8cm
SDP
Catalog No. 501367903
Encompass
Ideal for all staining, fixing and destaining procedures. Heavy duty staining dishes are molded from clear Pyrex brand borosilicate glass. Offers best compatibility with all chemical buffers and reagents. Safe to use with electrophoresis stains and dyes. Clean thoroughly after use. Supplied with tight fitting polyethylene lids to prevent evaporation of solutions.
Catalog No. 50-136-7903 Supplier Research Products International Corp Supplier No. 248933L
Ideal for all staining, fixing and destaining procedures. Heavy duty staining dishes are molded from clear Pyrex brand borosilicate glass. Offers best compatibility with all chemical buffers and reagents. Safe to use with electrophoresis stains and dyes. Clean thoroughly after use. Supplied with tight fitting polyethylene lids to prevent evaporation of solutions.