Learn More
Electron Microscopy Sciences Cryo Pucks, 12 indexed wells to hold round cryo grid boxes, Alpha-numeric code for easy identification, 24 grid boxes per puck
SDP
Catalog No. NC2734159
Encompass
Cryo Pucks provide storage and transport for electron microscopy samples on grids under cryogenic conditions. There are 12 indexed wells in each puck to hold round cryo grid boxes. The pucks have a unique alpha-numeric code for easy identification and sample tracking. The puck depth accomodates pin type lid style cryo grid boxes. When using cryo grid boxes with flat-style lids, you can store up to 24 cryo grid boxes per puck. Special tweezer slots allow easy and secure removal of cryo grid boxes.
Catalog No. NC2734159 Supplier Electron Microscopy Sciences Supplier No. 7116840
Cryo Pucks provide storage and transport for electron microscopy samples on grids under cryogenic conditions. There are 12 indexed wells in each puck to hold round cryo grid boxes. The pucks have a unique alpha-numeric code for easy identification and sample tracking. The puck depth accomodates pin type lid style cryo grid boxes. When using cryo grid boxes with flat-style lids, you can store up to 24 cryo grid boxes per puck. Special tweezer slots allow easy and secure removal of cryo grid boxes.