Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Pneumatic Spring, 130 lbs

Catalog No. NC1883822 Shop All Thermo Scientific Products
Encompass_Preferred

Pneumatic Spring, 130 lbs.

Catalog No. NC1883822 Supplier Thermo Scientific Supplier No. 129064
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pneumatic Spring, 130 lbs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.