Learn More
Chemglass Life Sciences 9-Neck PTFE Reaction Vessel Lid, 150mm, CN 45/50, SN 24/40 Outer, 112mm Overall Height
SDP
Catalog No. CG1945T74
Encompass_Preferred
9-Neck PTFE Reaction Vessel Lid, 150mm, CN 45/50, SN 24/40 Outer, 112mm Overall Height. Reaction vessel lid is constructed entirely of PTFE. Low-form lid has nine openings: (A) center neck 1 female NPT for PTFE standard taper adapter, (B) side necks 1 female NPT, vertical for PTFE standard taper adapters, (C) side neck 1 female NPT with 10 X 10 compound angle for PTFE compression fitting used with Lasentec Probe, and (D) side necks 1/2 female NPT. Lid is supplied complete with listed PTFE standard taper adapters for center and vertical side necks, 19mm PTFE compression fitting for 10 X 10 compound angle side neck, and four 1/2 male NPT PTFE plugs. For clamp see CG-141 or CG-141-T. For o-ring see CG-147. Special neck sizes and configurations are available. Please contact us for details.Lasentec is a registered trademark of Mettler Toledo.
Catalog No. CG1945T74 Supplier Chemglass Life Sciences Supplier No. CG1945T74
9-Neck PTFE Reaction Vessel Lid, 150mm, CN 45/50, SN 24/40 Outer, 112mm Overall Height. Reaction vessel lid is constructed entirely of PTFE. Low-form lid has nine openings: (A) center neck 1 female NPT for PTFE standard taper adapter, (B) side necks 1 female NPT, vertical for PTFE standard taper adapters, (C) side neck 1 female NPT with 10 X 10 compound angle for PTFE compression fitting used with Lasentec Probe, and (D) side necks 1/2 female NPT. Lid is supplied complete with listed PTFE standard taper adapters for center and vertical side necks, 19mm PTFE compression fitting for 10 X 10 compound angle side neck, and four 1/2 male NPT PTFE plugs. For clamp see CG-141 or CG-141-T. For o-ring see CG-147. Special neck sizes and configurations are available. Please contact us for details.Lasentec is a registered trademark of Mettler Toledo.