Learn More
Chemglass Life Sciences Reaction Vessel Lid, 5-Neck, 29/42 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange
SDP
Catalog No. 50873513
Encompass_Preferred
Reaction Vessel Lid, 5-Neck, 29/42 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.
Catalog No. 50-873-513 Supplier Chemglass Life Sciences Supplier No. CG194602
Reaction Vessel Lid, 5-Neck, 29/42 Center Neck, 4-24/40 Side Necks, 150mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.