Learn More
Chemglass Life Sciences 400mm Reaction Vessel Lid, 7-Neck, 45/50 CN, (5) 45/50 SNs, (1) 80mm O-ring Flange SN
SDP
Catalog No. 501531319
Encompass_Preferred
Reaction Vessel Lid, 7-neck, 400mm. Seven-Neck lid for 400mm flanged vessel. Lid has 45/50 center neck, five 45/50 side necks and a 80mm angled flange for powder addition. PLEASE NOTE: 80MM LID, O-RING AND CLAMP MUST BE ORDERED SEPARATELY.
Catalog No. 50-153-1319 Supplier Chemglass Life Sciences Supplier No. CG1968E400
Add to Cart
Add to Cart
Request a Quote
Reaction Vessel Lid, 7-neck, 400mm. Seven-Neck lid for 400mm flanged vessel. Lid has 45/50 center neck, five 45/50 side necks and a 80mm angled flange for powder addition. PLEASE NOTE: 80MM LID, O-RING AND CLAMP MUST BE ORDERED SEPARATELY.