Learn More
Chemglass Life Sciences Lid, Reaction Vessel, 5-Neck, 100mm Flange, 29/42 CN, 24/40 SN's: (2) Vertical and (2) Angled 10°
SDP
Catalog No. 50143904
Encompass_Preferred
Lid, Reaction Vessel, 5-Neck, 100mm Flange, 29/42 CN, 24/40SN's: (2) Vertical and (2) Angled 10¿. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Two side necks are vertical and two necks have a 10¿ degree angle. Angled side neck is designed for use with CG-1235/CG-1235-B series angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
Catalog No. 50-143-904 Supplier Chemglass Life Sciences Supplier No. CG1946D03
Lid, Reaction Vessel, 5-Neck, 100mm Flange, 29/42 CN, 24/40SN's: (2) Vertical and (2) Angled 10¿. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Two side necks are vertical and two necks have a 10¿ degree angle. Angled side neck is designed for use with CG-1235/CG-1235-B series angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.