Learn More
Chemglass Life Sciences Reaction Vessel Lid, 3-Neck, 45/50 Center Neck, 2-24/40 Side Necks, 150mm Schott Flange
SDP
Catalog No. 50873508
Encompass_Preferred
Reaction Vessel Lid, 3-Neck, 45/50 Center Neck, 2-24/40 Side Necks, 150mm Schott Flange. Three neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. NOTE: Three neck lids are available with joint sizes and configurations other than those listed, please call for details.
Catalog No. 50-873-508 Supplier Chemglass Life Sciences Supplier No. CG194006
Reaction Vessel Lid, 3-Neck, 45/50 Center Neck, 2-24/40 Side Necks, 150mm Schott Flange. Three neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. NOTE: Three neck lids are available with joint sizes and configurations other than those listed, please call for details.