Learn More
Chemglass Life Sciences Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-14/20 Side Necks on 30° angle, 60mm Schott Flange
SDP
Catalog No. 50873510
Encompass_Preferred
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-14/20 Side Necks on 30¿ angle, 60mm Schott Flange. Four neck reaction vessel lid to fit CG-1930 through CG-1931 vessels. Side necks are on a 30¿ angle. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
Catalog No. 50-873-510 Supplier Chemglass Life Sciences Supplier No. CG1943A01
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-14/20 Side Necks on 30¿ angle, 60mm Schott Flange. Four neck reaction vessel lid to fit CG-1930 through CG-1931 vessels. Side necks are on a 30¿ angle. Lid has a flat ground surface allowing it to be greased or used with an o-ring.