Learn More
Chemglass Life Sciences Lid, Reaction Vessel, 5-Neck, 100mm Flange ID, Low Profile, 112mm OAH, 29/42 CN, 24/40 SN's Angled 10°
SDP
Catalog No. 502733571
Encompass_Preferred
Low profile five neck vessel lid to fit CG-1920 through CG-1936 vessels. This low profile lid reduces overall height and provides less surface area for condensation. Two side necks are vertical and Two necks have a 10° degree angle. Angled side neck is designed for use with CG-1235/CG-1235-B series angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
Catalog No. 50-273-3571 Supplier Chemglass Life Sciences Supplier No. CG1946E03
Low profile five neck vessel lid to fit CG-1920 through CG-1936 vessels. This low profile lid reduces overall height and provides less surface area for condensation. Two side necks are vertical and Two necks have a 10° degree angle. Angled side neck is designed for use with CG-1235/CG-1235-B series angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.