Learn More
Chemglass Life Sciences Lid, Reaction Vessel, 5-Neck, 200mm Flange, 45/50 CN, (2) Vertical 45/50 Outer SNs, (2) 60mm O-Ring Flange SNs Angled 20
SDP
Catalog No. 502733572
Encompass_Preferred
This five-neck reaction vessel lid is designed for use with 200 mm flanged reaction vessels. It provides a central ground joint and multiple side necks (vertical and angled) to accommodate probes, condensers, and powder addition, and can be used with either a greased ground or an O-ring seal at the flange.
  • Five-neck configuration.
  • 200 mm flange.
  • Central 45/50 ground joint.
  • Two vertical 45/50 side necks.
  • Two 60 mm O-ring flange side necks angled 20 for powder addition.
  • Flat ground flange usable with grease or O-ring.
  • Fits 200 mm flanged reaction vessels.
Catalog No. 50-273-3572 Supplier Chemglass Life Sciences Supplier No. CG1946D50
This five-neck reaction vessel lid is designed for use with 200 mm flanged reaction vessels. It provides a central ground joint and multiple side necks (vertical and angled) to accommodate probes, condensers, and powder addition, and can be used with either a greased ground or an O-ring seal at the flange.
  • Five-neck configuration.
  • 200 mm flange.
  • Central 45/50 ground joint.
  • Two vertical 45/50 side necks.
  • Two 60 mm O-ring flange side necks angled 20 for powder addition.
  • Flat ground flange usable with grease or O-ring.
  • Fits 200 mm flanged reaction vessels.