Learn More
Chemglass Life Sciences Lid, Reaction Vessel, 5-Neck, 150mm Flange ID, Low Profile, 116mm OAH, 24/40 CN, 24/40 SN's Angled 10°
SDP
Catalog No. NC1316936
Encompass_Preferred
150mm Flange ID, Low Profile, 116mm OAH, 24/40 CN, 24/40 SN's Angled 10¿
Catalog No. NC1316936 Supplier Chemglass Life Sciences Supplier No. CG1946E05
150mm Flange ID, Low Profile, 116mm OAH, 24/40 CN, 24/40 SN's Angled 10¿