Learn More
Chemglass Life Sciences 5-Neck Reaction Vessel Lid, 150mm Schott Flange, CN 29/42, 3-SN 24/40 Vertical, 1-SN 24/40 Angled 10°
SDP
Catalog No. 501531287
Encompass_Preferred
5-Neck Reaction Vessel Lid, 150mm Schott Flange, CN 29/42, 3-SN 24/40 Vertical, 1-SN 24/40 Angled 10. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. four side necks are vertical and one neck has a 10 degree angle. Angled side neck is designed for use with CG-1235 angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
Catalog No. 50-153-1287 Supplier Chemglass Life Sciences Supplier No. CG1946B07
5-Neck Reaction Vessel Lid, 150mm Schott Flange, CN 29/42, 3-SN 24/40 Vertical, 1-SN 24/40 Angled 10. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. four side necks are vertical and one neck has a 10 degree angle. Angled side neck is designed for use with CG-1235 angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.